Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001764.2 | NP_001755.1 | 2 | 371 | ||
XM_011510119.2 | XP_011508421.1 | 2 | 878 | |||
XM_017002784.1 | XP_016858273.1 | 2 | 878 | |||
XM_017002785.1 | XP_016858274.1 | 2 | 878 | |||
XM_017002786.1 | XP_016858275.1 | 2 | 878 | |||
GenBank mRNA | AK303329.1 | - | 2 | 686 | ||
AK303330.1 | - | 2 | 278 | |||
AK310617.1 | - | 2 | 288 | |||
BC069481.1 | - | 2 | 308 | 7262331 | ||
BC074747.2 | - | 2 | 285 | 30915207 | ||
BC104216.1 | - | 2 | 308 | 40031984 | ||
BC104217.1 | - | 2 | 308 | 40031985 | ||
CD687240.1 | - | 2 | 401 | |||
M28826.1 | - | 2 | 308 |