Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003058.3 | NP_003049.2 | 11 | 2432 | ||
GenBank mRNA | AB075951.1 | - | 10 | 3605 | ||
BC039899.1 | - | 11 | 2310 | 5186229 |