Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 17 | 2115 | ||
XM_011540326.1 | XP_011538628.1 | 17 | 2214 | |||
XM_011540327.2 | XP_011538629.1 | 14 | 1880 | |||
XM_011540328.2 | XP_011538630.1 | 14 | 1708 | |||
XM_011540329.2 | XP_011538631.1 | 17 | 2325 | |||
XM_017016877.1 | XP_016872366.1 | 16 | 2177 | |||
XM_017016878.1 | XP_016872367.1 | 16 | 2007 | |||
XM_017016879.1 | XP_016872368.1 | 16 | 2131 | |||
XM_017016880.1 | XP_016872369.1 | 16 | 2135 | |||
XM_017016881.1 | XP_016872370.1 | 15 | 1984 | |||
GenBank mRNA | AB208843.1 | - | 4 | 404 | ||
AF038362.1 | - | 17 | 2134 | |||
AJ001017.2 | - | 17 | 2029 | |||
BC030273.1 | - | 11 | 1362 | |||
BC047009.2 | - | 5 | 607 | |||
BC112201.1 | - | 17 | 2099 | 8327669 |