Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004799.3 | NP_004790.2 | 4 | 1512 | ||
NM_007324.3 | NP_015563.2 | 4 | 1512 | |||
XM_011542437.2 | XP_011540739.1 | 4 | 1383 | |||
XM_017002845.1 | XP_016858334.1 | 4 | 1605 | |||
GenBank mRNA | AB209125.1 | - | 1 | 786 | ||
AF104304.1 | - | 3 | 1192 | |||
AF130419.1 | - | 4 | 1232 | |||
AF130420.1 | - | 4 | 1232 | |||
BC032680.1 | - | 2 | 1071 | 5585817 |