Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE1 SIRNA Assays ›
Gene Symbol:
TLE1
Gene Name:
transducin like enhancer of split 1
Gene Aliases:
ESG, ESG1, GRG1
Chromosome Location:
Chr.9: 81583683 - 81689535 on Build GRCh38
UniGene ID:
Hs.197320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107627
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303103.1
NP_001290032.1
11
2176
NM_001303104.1
NP_001290033.1
11
2146
NM_005077.4
NP_005068.2
11
2146
XM_005252151.1
XP_005252208.1
11
1090
XM_005252152.1
XP_005252209.1
11
1090
XM_005252153.1
XP_005252210.1
11
1060
XM_005252154.1
XP_005252211.1
11
1057
XM_005252156.2
XP_005252213.1
11
1090
XM_005252162.1
XP_005252219.1
9
838
XM_005252163.2
XP_005252220.1
9
838
XM_006717258.1
XP_006717321.1
11
1087
XM_006717259.3
XP_006717322.1
9
773
XM_006717260.1
XP_006717323.1
9
868
XM_006717261.2
XP_006717324.1
9
802
XM_006717262.1
XP_006717325.1
11
1060
XM_006717263.1
XP_006717326.1
11
1057
XM_011518951.2
XP_011517253.1
11
1057
XM_017015064.1
XP_016870553.1
11
1087
XM_017015065.1
XP_016870554.1
11
1087
XM_017015066.1
XP_016870555.1
9
830
GenBank mRNA
AB209854.1
-
11
2146
AK290860.1
-
11
908
AK293606.1
-
11
934
AK304565.1
-
9
747
BC010100.2
-
11
1292
3506451
BC015747.1
-
11
1002
4857394
M99435.1
-
11
876
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
multicellular organism development
organ morphogenesis
positive regulation of gene expression
Wnt signaling pathway
negative regulation of Wnt signaling pathway
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of transcription, DNA-templated
beta-catenin-TCF complex assembly
negative regulation of anoikis
RNA polymerase II transcription corepressor activity
protein binding
transcription factor binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0