Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005415.4 | NP_005406.3 | 11 | 3101 | ||
XM_017004768.1 | XP_016860257.1 | 11 | 2850 | |||
XM_017004769.1 | XP_016860258.1 | 12 | 5328 | |||
XM_017004770.1 | XP_016860259.1 | 12 | 3131 | |||
XM_017004771.1 | XP_016860260.1 | 13 | 3167 | |||
GenBank mRNA | AK123420.1 | - | 5 | 2138 | ||
BC019944.1 | - | 11 | 3008 | 3918690 | ||
BC075818.1 | - | 5 | 1610 | |||
BX647555.1 | - | 3 | 2930 | |||
L20859.1 | - | 11 | 2931 |