Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 8 | 2390 | ||
NM_006074.4 | NP_006065.2 | 8 | 2402 | |||
GenBank mRNA | AL360190.1 | - | 1 | 813 | ||
AM040752.1 | - | 5 | 1350 | |||
BC022281.1 | - | 8 | 2210 | 4718024 | ||
BC035582.1 | - | 8 | 2175 | 5583800 |