Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127370.2 | NP_001120842.1 | 4 | 438 | ||
NM_018719.4 | NP_061189.2 | 3 | 384 | |||
GenBank mRNA | AK022955.1 | - | 3 | 359 | ||
AK095965.1 | - | 4 | 392 | |||
AK292490.1 | - | 3 | 359 | |||
AY161168.1 | - | 3 | 261 | |||
AY161169.1 | - | 3 | 264 | |||
BC009352.2 | - | 3 | 276 | 4139111 | ||
BC014630.2 | - | 2 | 228 | |||
BC025242.1 | - | 3 | 338 | 4842169 | ||
BC032576.1 | - | 3 | 349 | 5491780 | ||
BC039823.1 | - | 3 | 348 | 6057601 | ||
DA041070.1 | - | 3 | 384 |