Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174089.1 | NP_001167560.1 | 20 | 3059 | ||
NM_001174090.1 | NP_001167561.1 | 20 | 3209 | |||
NM_032034.3 | NP_114423.1 | 19 | 3051 | |||
NR_135000.1 | 20 | 3053 | ||||
XM_005260856.4 | XP_005260913.1 | 19 | 3433 | |||
XM_005260857.1 | XP_005260914.1 | 20 | 3078 | |||
XM_011529383.2 | XP_011527685.1 | 20 | 4654 | |||
XM_011529384.1 | XP_011527686.1 | 20 | 3296 | |||
XM_011529385.1 | XP_011527687.1 | 20 | 3123 | |||
XM_017028093.1 | XP_016883582.1 | 20 | 3427 | |||
XM_017028094.1 | XP_016883583.1 | 20 | 2931 | |||
XM_017028095.1 | XP_016883584.1 | 19 | 3288 | |||
XM_017028096.1 | XP_016883585.1 | 21 | 3308 | |||
GenBank mRNA | AF336127.1 | - | 19 | 3051 | ||
AK075303.1 | - | 14 | 2326 | |||
AK296508.1 | - | 20 | 3209 |