Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145283.2 | NP_660326.2 | 3 | 1148 | ||
XM_005251727.3 | XP_005251784.1 | 2 | 1032 | |||
GenBank mRNA | BC022521.1 | - | 3 | 1144 | 4793993 |