Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110427
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204148.2
NP_001191077.1
6
907
NM_001321869.1
NP_001308798.1
8
1189
NM_001321870.1
NP_001308799.1
8
1180
NM_001321874.1
NP_001308803.1
7
1110
NM_001321875.1
NP_001308804.1
7
1116
NM_001321876.1
NP_001308805.1
7
1104
NM_001321878.1
NP_001308807.1
7
1101
NM_001321884.1
NP_001308813.1
7
1104
NM_001321886.1
NP_001308815.1
8
1160
NM_001321887.1
NP_001308816.1
8
994
NM_001321890.1
NP_001308819.1
7
1095
NM_001321896.1
NP_001308825.1
8
1279
NM_001321899.1
NP_001308828.1
8
1007
NM_001321900.1
NP_001308829.1
7
1101
NM_001321901.1
NP_001308830.1
7
935
NM_001321902.1
NP_001308831.1
7
941
NM_014707.3
NP_055522.1
7
932
NM_058176.2
NP_478056.1
6
787
NM_178423.2
NP_848510.1
7
932
NM_178425.3
NP_848512.1
6
796
XM_011515629.2
XP_011513931.1
7
890
XM_011515631.2
XP_011513933.1
8
1204
XM_011515635.2
XP_011513937.1
8
1756
XM_011515636.2
XP_011513938.1
7
1477
XM_011515637.2
XP_011513939.1
7
901
XM_011515638.2
XP_011513940.1
7
1471
XM_011515639.2
XP_011513941.1
7
1668
XM_011515641.2
XP_011513943.1
8
1753
XM_011515642.2
XP_011513944.1
6
2482
XM_011515644.2
XP_011513946.1
8
1728
XM_011515645.2
XP_011513947.1
6
2467
XM_011515646.2
XP_011513948.1
6
2458
XM_011515648.2
XP_011513950.1
7
1486
XM_011515649.2
XP_011513951.1
7
1690
XM_017012826.1
XP_016868315.1
7
1462
XM_017012827.1
XP_016868316.1
8
1545
XM_017012829.1
XP_016868318.1
7
865
XM_017012830.1
XP_016868319.1
7
856
XM_017012832.1
XP_016868321.1
7
865
GenBank mRNA
AB018287.1
-
7
896
AF124924.1
-
4
454
AJ459808.1
-
7
896
AK304343.1
-
6
827
AY032737.1
-
6
746
AY032738.1
-
6
746
AY197371.1
-
6
755
BC150328.1
-
7
907
8860495
BC152405.1
-
7
896
8862508
CR749550.1
-
6
863
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0