Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001263.3 | NP_001254.2 | 13 | 1809 | ||
XM_005262687.2 | XP_005262744.1 | 12 | 1685 | |||
XM_017007648.1 | XP_016863137.1 | 12 | 1736 | |||
XM_017007649.1 | XP_016863138.1 | 11 | 1616 | |||
XM_017007650.1 | XP_016863139.1 | 11 | 1581 | |||
XM_017007651.1 | XP_016863140.1 | 10 | 1512 | |||
GenBank mRNA | AK314245.1 | - | 13 | 1548 | ||
BC074833.2 | - | 13 | 1360 | 30915545 | ||
BC074881.2 | - | 13 | 1360 | 30915239 | ||
U60808.1 | - | 13 | 1460 | |||
U65887.1 | - | 13 | 1747 |