Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191002.1 | NP_001177931.1 | 3 | 404 | ||
NM_001191003.1 | NP_001177932.1 | 3 | 457 | |||
NM_004832.2 | NP_004823.1 | 3 | 404 | |||
GenBank mRNA | AF212303.1 | - | 3 | 211 | ||
AK313680.1 | - | 3 | 222 | |||
BC000127.1 | - | 3 | 214 | 3351359 | ||
BX537431.1 | - | 3 | 332 | |||
CF130513.1 | - | 3 | 181 | |||
CN283947.1 | - | 3 | 207 | |||
DT220068.1 | - | 3 | 439 | |||
EU794619.1 | - | 3 | 211 | |||
U90313.1 | - | 3 | 194 |