Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284367.1 | NP_001271296.1 | 5 | 762 | ||
NM_001284368.1 | NP_001271297.1 | 5 | 461 | |||
NM_005004.3 | NP_004995.1 | 5 | 552 | |||
GenBank mRNA | AF044958.1 | - | 5 | 502 | ||
AF115968.1 | - | 5 | 508 | |||
AK289579.1 | - | 5 | 493 | |||
AL080056.1 | - | 5 | 512 | |||
BC000466.2 | - | 5 | 493 | 2822781 | ||
BC019276.1 | - | 5 | 493 | 2822781 | ||
BM008944.1 | - | 5 | 449 | |||
CF127298.1 | - | 5 | 491 | |||
CR533490.1 | - | 5 | 480 |