Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083615.3 | NP_001077084.2 | 34 | 5045 | ||
NM_138995.4 | NP_620482.3 | 35 | 5126 | |||
NR_045682.1 | 36 | 5248 | ||||
NR_045683.1 | 36 | 5157 | ||||
NR_045684.1 | 35 | 5076 | ||||
XM_006712299.3 | XP_006712362.1 | 35 | 5014 | |||
XM_011510654.2 | XP_011508956.1 | 36 | 5122 | |||
XM_011510655.2 | XP_011508957.1 | 34 | 4933 | |||
XM_011510656.2 | XP_011508958.1 | 30 | 4497 | |||
XM_011510657.2 | XP_011508959.1 | 29 | 4369 | |||
XM_011510658.2 | XP_011508960.1 | 30 | 4678 | |||
GenBank mRNA | AF369908.1 | - | 36 | 5157 | ||
AF391554.1 | - | 35 | 5076 | |||
AF391555.1 | - | 35 | 5126 | |||
AF391556.1 | - | 34 | 5045 | |||
BX647895.1 | - | 28 | 4211 |