Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173483.3 | NP_775754.2 | 14 | 2578 | ||
XM_011527692.2 | XP_011525994.1 | 15 | 2820 | |||
XM_011527693.2 | XP_011525995.1 | 14 | 2590 | |||
GenBank mRNA | AK096820.1 | - | 14 | 2578 |