Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP8 SIRNA Assays ›
Gene Symbol:
DPP8
Gene Name:
dipeptidyl peptidase 8
Gene Aliases:
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location:
Chr.15: 65442463 - 65517713 on Build GRCh38
UniGene ID:
Hs.458609
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114004
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320875.1
NP_001307804.1
3
482
NM_001320876.1
NP_001307805.1
2
451
NM_017743.5
NP_060213.2
2
346
NM_130434.4
NP_569118.1
2
346
NM_197960.3
NP_932064.1
3
387
NM_197961.3
NP_932065.1
3
482
NR_135485.1
2
346
NR_135486.1
3
387
XM_011521730.1
XP_011520032.1
2
1735
XM_011521731.1
XP_011520033.1
2
1735
XM_011521732.2
XP_011520034.1
2
1735
XM_011521733.1
XP_011520035.1
2
327
XM_011521734.1
XP_011520036.1
2
1735
XM_017022373.1
XP_016877862.1
2
1735
XM_017022374.1
XP_016877863.1
2
1735
XM_017022375.1
XP_016877864.1
2
327
XM_017022376.1
XP_016877865.1
2
327
XM_017022377.1
XP_016877866.1
2
327
XM_017022378.1
XP_016877867.1
2
1735
XM_017022379.1
XP_016877868.1
2
327
XM_017022380.1
XP_016877869.1
2
327
XM_017022381.1
XP_016877870.1
2
327
GenBank mRNA
AF221634.1
-
2
320
AK000290.1
-
2
269
AK291057.1
-
3
457
AK296915.1
-
2
422
AK302638.1
-
3
323
AY172659.1
-
1
114
AY354202.1
-
3
315
BC030688.2
-
3
473
4822550
BI462092.1
-
3
328
DA297112.1
-
2
332
DC401507.1
-
3
388
DC412877.1
-
2
348
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
immune response
serine-type peptidase activity
dipeptidyl-peptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0