Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267868.1 | NP_001254797.1 | 6, 7 | 1075 | ||
NM_001267869.1 | NP_001254798.1 | 3, 4 | 762 | |||
NM_001267870.1 | NP_001254799.1 | 2, 3 | 602 | |||
NM_001267871.1 | NP_001254800.1 | 3, 4 | 694 | |||
NM_198533.2 | NP_940935.1 | 5, 6 | 937 | |||
NM_198704.2 | NP_941993.1 | 3, 4 | 762 | |||
NM_198705.2 | NP_941994.1 | 3, 4 | 694 | |||
NM_198706.2 | NP_941995.1 | 5, 6 | 937 | |||
NM_198707.2 | NP_941996.1 | 4, 5 | 874 | |||
NM_198708.2 | NP_941997.1 | 4, 5 | 825 | |||
GenBank mRNA | AK308206.1 | - | 6, 7 | 815 | ||
AY268351.1 | - | 3, 4 | 435 | |||
AY268352.1 | - | 5, 6 | 638 | |||
AY268353.1 | - | 4, 5 | 598 | |||
AY268354.1 | - | 4, 5 | 526 | |||
AY313894.1 | - | 5, 6 | 666 | |||
AY313895.1 | - | 3, 4 | 503 | |||
AY313896.1 | - | 3, 4 | 435 | |||
BC053546.1 | - | 4, 5 | 455 | |||
BC072434.1 | - | 3, 4 | 511 | |||
BC093950.1 | - | 1, 2 | 101 | |||
BC094798.1 | - | 3, 4 | 518 | |||
BC112187.1 | - | 1, 2 | 101 | |||
CX865680.1 | - | 2, 3 | 208 |