Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000523.3 | NP_000514.2 | 2 | 1037 | ||
XM_011511068.2 | XP_011509370.1 | 2 | 1348 | |||
GenBank mRNA | BG204577.1 | - | 1 | 173 |