Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261435.1 | NP_001248364.1 | 14 | 1547 | ||
NM_139158.2 | NP_631897.1 | 14 | 1475 | |||
XM_005246781.3 | XP_005246838.1 | 14 | 1565 | |||
XM_005246782.4 | XP_005246839.1 | 15 | 1812 | |||
XM_011511652.1 | XP_011509954.1 | 12 | 1376 | |||
XM_011511655.2 | XP_011509957.1 | 11 | 1572 | |||
XM_011511656.2 | XP_011509958.1 | 11 | 1179 | |||
XM_011511657.2 | XP_011509959.1 | 9 | 1076 | |||
GenBank mRNA | AB053308.1 | - | 14 | 1429 | ||
AK091710.1 | - | 4 | 569 | |||
AK292434.1 | - | 15 | 1682 | |||
AX772791.1 | - | 14 | 1429 | |||
BX647394.1 | - | 14 | 1510 |