Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000280.4 | NP_000271.1 | 13 | 1797 | ||
NM_001127612.1 | NP_001121084.1 | 13 | 1711 | |||
NM_001258462.1 | NP_001245391.1 | 14 | 1753 | |||
NM_001258463.1 | NP_001245392.1 | 14 | 1691 | |||
NM_001258464.1 | NP_001245393.1 | 13 | 1699 | |||
NM_001258465.1 | NP_001245394.1 | 12 | 1685 | |||
NM_001310158.1 | NP_001297087.1 | 14 | 1794 | |||
NM_001310160.1 | NP_001297089.1 | 10 | 3008 | |||
NM_001310161.1 | NP_001297090.1 | 11 | 1560 | |||
NM_001604.5 | NP_001595.2 | 14 | 1741 | |||
GenBank mRNA | AB209177.1 | - | 11 | 6149 | ||
AB593092.1 | - | 14 | 1692 | |||
AB593093.1 | - | 14 | 1691 | |||
AB593094.1 | - | 14 | 1794 | |||
AK074881.1 | - | 12 | 2132 | |||
AK094172.1 | - | 8 | 2435 | |||
AK094249.1 | - | 10 | 2818 | |||
AK314470.1 | - | 13 | 1689 | |||
AY047583.1 | - | 11 | 1372 | |||
AY707088.1 | - | 9 | 1242 | |||
BC011953.1 | - | 13 | 1583 | 3880468 | ||
BI816814.1 | - | 4 | 465 | |||
BX640762.1 | - | 14 | 1720 | |||
CV569250.1 | - | 2 | 221 | |||
M77844.1 | - | 12 | 1604 | |||
M93650.1 | - | 13 | 1659 |