Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244682.1 | NP_001231611.1 | 13 | 1362 | ||
NM_014352.3 | NP_055167.2 | 13 | 1472 | |||
XM_011542739.2 | XP_011541041.1 | 15 | 1687 | |||
XM_011542740.2 | XP_011541042.1 | 14 | 1545 | |||
XM_011542741.2 | XP_011541043.1 | 12 | 1396 | |||
XM_011542742.2 | XP_011541044.1 | 14 | 1650 | |||
XM_011542743.2 | XP_011541045.1 | 12 | 3791 | |||
GenBank mRNA | AF133895.1 | - | 13 | 1329 | ||
AF162278.1 | - | 13 | 1436 | |||
AJ012214.1 | - | 12 | 1322 | |||
AK097382.1 | - | 6 | 851 | |||
AK291993.1 | - | 13 | 1472 | |||
AK302212.1 | - | 13 | 1362 | |||
BC101649.1 | - | 13 | 1407 | 8069155 | ||
BC143593.1 | - | 14 | 1454 | |||
BC143594.1 | - | 14 | 1451 | |||
BC143595.1 | - | 14 | 1452 |