Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130441.2 | NP_569708.1 | 7 | 1257 | ||
NM_203503.1 | NP_987099.1 | 6 | 1164 | |||
XM_017018940.1 | XP_016874429.1 | 6 | 912 | |||
GenBank mRNA | AF293615.1 | - | 7 | 1257 | ||
AK122596.1 | - | 6 | 940 | |||
AY358223.1 | - | 6 | 923 |