siRNA Details
Gene Symbol: SOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003107.2 | NP_003098.1 | 1 | 2550 | ||
GenBank mRNA | CN419098.1 | - | 1 | 204 | ||
X70683.1 | - | 1 | 2104 |

Gene Symbol: SOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003107.2 | NP_003098.1 | 1 | 2550 | ||
GenBank mRNA | CN419098.1 | - | 1 | 204 | ||
X70683.1 | - | 1 | 2104 |