Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017371.4 | NP_001017371.3 | 5 | 2427 | ||
NM_001172712.1 | NP_001166183.1 | 7 | 2769 | |||
NM_003111.4 | NP_003102.1 | 7 | 2778 | |||
GenBank mRNA | AB209334.1 | - | 4 | 1891 | ||
AK304199.1 | - | 7 | 2350 | |||
AY070137.1 | - | 5 | 2427 | |||
AY441957.1 | - | 7 | 2778 | |||
AY441958.1 | - | 5 | 2117 | |||
BC042945.2 | - | 4 | 981 | |||
BC126414.1 | - | 7 | 2300 | 8992130 | ||
BC143928.1 | - | 7 | 2291 | 9052446 | ||
CR749323.1 | - | 2 | 3984 | |||
M97191.2 | - | 5 | 2038 | |||
X68560.1 | - | 5 | 1996 |