Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MECOM SIRNA Assays ›
Gene Symbol:
MECOM
Gene Name:
MDS1 and EVI1 complex locus
Gene Aliases:
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location:
Chr.3: 169083499 - 169663781 on Build GRCh38
UniGene ID:
Hs.744090
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115628
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MECOM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105077.3
NP_001098547.3
5
516
NM_001105078.3
NP_001098548.2
4
510
NM_001163999.1
NP_001157471.1
4
454
NM_001164000.1
NP_001157472.1
4
1386
NM_001205194.1
NP_001192123.1
4
814
NM_004991.3
NP_004982.2
5
1152
NM_005241.3
NP_005232.2
4
454
XM_005247213.3
XP_005247270.1
5
1158
XM_005247214.3
XP_005247271.1
5
1158
XM_005247215.3
XP_005247272.1
5
1157
XM_005247219.2
XP_005247276.1
4
1123
XM_005247220.2
XP_005247277.1
4
464
XM_005247221.2
XP_005247278.1
4
447
XM_005247223.2
XP_005247280.1
4
1123
XM_005247224.3
XP_005247281.1
5
1158
XM_005247225.3
XP_005247282.1
5
1157
XM_005247226.3
XP_005247283.1
5
1035
XM_011512546.2
XP_011510848.1
4
911
XM_017005874.1
XP_016861363.1
4
1826
XM_017005875.1
XP_016861364.1
4
464
XM_017005876.1
XP_016861365.1
4
1123
XM_017005877.1
XP_016861366.1
5
1157
XM_017005878.1
XP_016861367.1
4
1123
GenBank mRNA
AF164154.1
-
4
1655
AF164155.1
-
5
1410
AF164156.1
-
5
971
AF164157.1
-
5
498
AK292865.1
-
4
437
AK304098.1
-
5
1098
AK308404.1
-
5
797
AK310509.1
-
3
298
BC130520.1
-
5
490
40146551
BC143951.1
-
5
490
9052469
BC143952.1
-
4
428
9052470
BX640908.1
-
4
1386
BX647613.1
-
4
1118
GQ352634.1
-
4
718
X54989.1
-
4
452
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
cell differentiation
histone lysine methylation
negative regulation of programmed cell death
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of JNK cascade
regulation of cell cycle
hematopoietic stem cell proliferation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0