Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278119.1 | NP_001265048.1 | 3 | 516 | ||
NM_001278121.1 | NP_001265050.1 | 3 | 449 | |||
NM_001278122.1 | NP_001265051.1 | 2 | 422 | |||
NM_006298.3 | NP_006289.2 | 2 | 422 | |||
XM_011514870.2 | XP_011513172.1 | 2 | 511 | |||
XM_011514873.2 | XP_011513175.1 | 3 | 608 | |||
XM_017011266.1 | XP_016866755.1 | 2 | 426 | |||
GenBank mRNA | AA453245.1 | - | 1 | 69 | ||
AB208793.1 | - | 2 | 422 | |||
AK302402.1 | - | 3 | 449 | |||
BC096282.3 | - | 3 | 488 | 40006900 | ||
BC096283.3 | - | 2 | 394 | 40006903 | ||
BC096284.3 | - | 2 | 394 | 40006905 | ||
BC113961.1 | - | 2 | 431 | 40022943 | ||
BC130032.1 | - | 2 | 412 | 40018337 | ||
BC152378.1 | - | 2 | 431 | 40018341 | ||
BQ327604.1 | - | 2 | 424 | |||
U57796.1 | - | 2 | 394 |