Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOR1 SIRNA Assays ›
Gene Symbol:
NCOR1
Gene Name:
nuclear receptor corepressor 1
Gene Aliases:
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location:
Chr.17: 16030094 - 16216746 on Build GRCh38
UniGene ID:
Hs.462323
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115788
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190438.1
NP_001177367.1
17
2292
NM_001190440.1
NP_001177369.1
19
2619
NM_006311.3
NP_006302.2
20
2770
XM_005256866.4
XP_005256923.1
21
2831
XM_005256868.4
XP_005256925.1
21
2828
XM_005256871.4
XP_005256928.1
20
2804
XM_005256872.4
XP_005256929.1
20
2801
XM_005256873.4
XP_005256930.1
21
2783
XM_005256874.4
XP_005256931.1
21
2831
XM_005256875.3
XP_005256932.1
21
2831
XM_006721601.3
XP_006721664.1
22
2963
XM_006721602.3
XP_006721665.1
22
2958
XM_006721603.3
XP_006721666.1
21
2831
XM_006721604.3
XP_006721667.1
20
2801
XM_011524083.2
XP_011522385.1
21
3742
XM_011524084.2
XP_011522386.1
21
2831
XM_011524085.2
XP_011522387.1
21
2831
XM_011524086.2
XP_011522388.1
20
2801
XM_017025396.1
XP_016880885.1
22
3708
XM_017025397.1
XP_016880886.1
22
3473
XM_017025398.1
XP_016880887.1
21
2915
XM_017025399.1
XP_016880888.1
21
2934
XM_017025400.1
XP_016880889.1
22
3705
XM_017025401.1
XP_016880890.1
22
3470
XM_017025402.1
XP_016880891.1
21
2754
XM_017025403.1
XP_016880892.1
21
2931
XM_017025404.1
XP_016880893.1
21
3422
XM_017025405.1
XP_016880894.1
20
3317
XM_017025406.1
XP_016880895.1
20
3715
XM_017025407.1
XP_016880896.1
20
2651
XM_017025408.1
XP_016880897.1
20
2723
XM_017025409.1
XP_016880898.1
21
2928
XM_017025410.1
XP_016880899.1
21
3424
XM_017025411.1
XP_016880900.1
20
3167
XM_017025412.1
XP_016880901.1
20
2944
XM_017025413.1
XP_016880902.1
20
2727
XM_017025414.1
XP_016880903.1
19
2634
XM_017025415.1
XP_016880904.1
21
2831
XM_017025416.1
XP_016880905.1
21
3439
XM_017025417.1
XP_016880906.1
21
2783
XM_017025418.1
XP_016880907.1
21
2831
XM_017025419.1
XP_016880908.1
21
2831
XM_017025420.1
XP_016880909.1
21
2783
GenBank mRNA
AB028970.2
-
19
2593
AF044209.1
-
20
2741
AF087856.1
-
17
2306
AF303586.1
-
17
2306
BC167431.1
-
19
2594
8860107
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
skeletal muscle tissue development
lactation
circadian rhythm
positive regulation of cell proliferation
covalent chromatin modification
cerebellum development
cellular lipid metabolic process
negative regulation of JNK cascade
spindle assembly
regulation of glycolytic process by negative regulation of transcription from RNA polymerase II promoter
regulation of lipid transport by negative regulation of transcription from RNA polymerase II promoter
negative regulation of production of miRNAs involved in gene silencing by miRNA
regulation of fatty acid transport
RNA polymerase II regulatory region DNA binding
RNA polymerase II activating transcription factor binding
chromatin binding
transcription corepressor activity
protein binding
ligand-dependent nuclear receptor binding
protein domain specific binding
estrogen receptor binding
nuclear hormone receptor binding
histone deacetylase binding
peroxisome proliferator activated receptor binding
transcription regulatory region DNA binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0