Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006841.5 | NP_006832.1 | 8 | 726 | ||
XM_006712954.2 | XP_006713017.1 | 8 | 760 | |||
GenBank mRNA | AF244548.1 | - | 8 | 685 | ||
AK293801.1 | - | 7 | 513 | |||
AK299677.1 | - | 7 | 630 | |||
AK313461.1 | - | 8 | 700 | |||
BC042875.1 | - | 8 | 698 | 5174617 | ||
BX537382.1 | - | 8 | 700 | |||
CR456970.1 | - | 7 | 572 | |||
U49082.1 | - | 8 | 696 |