Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006933.6 | NP_008864.4 | 2 | 912 | ||
GenBank mRNA | AX750578.1 | - | 2 | 865 |