Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100876.1 | NP_001094346.1 | 5 | 1156 | ||
NM_001100877.1 | NP_001094347.1 | 3 | 264 | |||
NM_174933.3 | NP_777593.2 | 5 | 1156 | |||
GenBank mRNA | AK054994.1 | - | 6 | 668 | ||
AK095000.1 | - | 5 | 1156 | |||
BC009373.2 | - | 5 | 643 | 4121964 | ||
BC051300.1 | - | 3 | 236 | 4843267 | ||
BC053853.1 | - | 3 | 225 | 5209351 |