Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287498.1 | NP_001274427.1 | 9 | 1005 | ||
NM_032295.3 | NP_115671.1 | 9 | 1005 | |||
NM_207113.2 | NP_996996.1 | 9 | 1005 | |||
XM_011516626.2 | XP_011514928.1 | 11 | 1502 | |||
XM_011516627.2 | XP_011514929.1 | 11 | 1501 | |||
XM_017012712.1 | XP_016868201.1 | 11 | 1478 | |||
XM_017012713.1 | XP_016868202.1 | 11 | 1497 | |||
XM_017012714.1 | XP_016868203.1 | 9 | 1275 | |||
XM_017012717.1 | XP_016868206.1 | 11 | 1497 | |||
GenBank mRNA | AK074823.1 | - | 9 | 966 | ||
AK299276.1 | - | 10 | 1051 | |||
AK300265.1 | - | 8 | 1335 | |||
AL136583.1 | - | 9 | 912 | |||
BC028380.1 | - | 9 | 955 | 4811814 | ||
BC046567.1 | - | 9 | 957 | 5246993 |