Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004523.3 | NP_004514.2 | 1, 2 | 395 | ||
GenBank mRNA | AK314258.1 | - | 1, 2 | 298 | ||
BC126211.1 | - | 1, 2 | 194 | 8991927 | ||
BC136474.1 | - | 1, 2 | 194 | 9020463 | ||
DB078134.1 | - | 1, 2 | 395 | |||
U37426.1 | - | 1, 2 | 177 | |||
X85137.1 | - | 1, 2 | 97 |