Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDKL3 SIRNA Assays ›
Gene Symbol:
CDKL3
Gene Name:
cyclin dependent kinase like 3
Gene Aliases:
NKIAMRE
Chromosome Location:
Chr.5: 134298424 - 134371159 on Build GRCh38
UniGene ID:
Hs.719926
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118806
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDKL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113575.1
NP_001107047.1
9
1421
NM_001300853.1
NP_001287782.1
8
1187
NM_016508.3
NP_057592.2
9
1421
XM_011543429.2
XP_011541731.1
9
1772
XM_011543439.2
XP_011541741.1
6
925
XM_011543444.2
XP_011541746.1
5
754
XM_011543445.2
XP_011541747.1
5
703
XM_011543447.2
XP_011541749.1
4
614
XM_017009524.1
XP_016865013.1
9
2249
XM_017009525.1
XP_016865014.1
10
1817
XM_017009526.1
XP_016865015.1
10
1815
XM_017009527.1
XP_016865016.1
9
2213
XM_017009528.1
XP_016865017.1
8
2204
XM_017009529.1
XP_016865018.1
9
2249
XM_017009530.1
XP_016865019.1
9
2249
XM_017009531.1
XP_016865020.1
9
2249
XM_017009532.1
XP_016865021.1
8
2167
XM_017009533.1
XP_016865022.1
8
2203
XM_017009534.1
XP_016865023.1
8
2203
XM_017009535.1
XP_016865024.1
8
2167
XM_017009536.1
XP_016865025.1
6
1962
XM_017009537.1
XP_016865026.1
7
1367
XM_017009538.1
XP_016865027.1
9
1257
XM_017009539.1
XP_016865028.1
8
1078
XM_017009540.1
XP_016865029.1
7
888
XM_017009541.1
XP_016865030.1
8
1414
XM_017009542.1
XP_016865031.1
7
845
XM_017009543.1
XP_016865032.1
7
872
XM_017009544.1
XP_016865033.1
7
1123
XM_017009545.1
XP_016865034.1
6
930
XM_017009546.1
XP_016865035.1
6
959
XM_017009547.1
XP_016865036.1
4
651
XM_017009548.1
XP_016865037.1
7
2054
GenBank mRNA
AF130372.1
-
8
1302
AK294559.1
-
6
809
AK294740.1
-
5
719
AK297586.1
-
4
581
AK299305.1
-
5
669
AK301802.1
-
8
1181
AK310955.1
-
5
659
BC041799.1
-
9
1423
5298279
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein phosphorylation
regulation of cell cycle
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0