Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303419.1 | NP_001290348.1 | 2 | 465 | ||
NM_173553.2 | NP_775824.2 | 2 | 465 | |||
XM_011531737.2 | XP_011530039.1 | 3 | 656 | |||
XM_011531738.1 | XP_011530040.1 | 3 | 826 | |||
XM_011531739.1 | XP_011530041.1 | 1 | 45 | |||
XM_017007882.1 | XP_016863371.1 | 3 | 511 | |||
GenBank mRNA | AK098667.1 | - | 2 | 132 | ||
AK300428.1 | - | 3 | 494 | |||
BC111959.1 | - | 2 | 126 | 8327427 | ||
BC111961.1 | - | 2 | 126 | 8327429 | ||
BC143709.1 | - | 2 | 126 | 9052223 |