Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290042.1 | NP_001276971.1 | 2 | 328 | ||
NM_002783.2 | NP_002774.2 | 2 | 328 | |||
GenBank mRNA | BC030979.1 | - | 2 | 323 | ||
BG434892.1 | - | 2 | 325 | |||
BX952498.1 | - | 2 | 326 | |||
U18467.1 | - | 2 | 278 |