Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119963
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
5
855
NM_001258380.1
NP_001245309.1
5
617
NM_001321103.1
NP_001308032.1
5
867
NM_001321104.1
NP_001308033.1
5
867
NM_001321105.1
NP_001308034.1
5
867
NM_001321106.1
NP_001308035.1
5
855
NM_001321107.1
NP_001308036.1
5
855
NM_001321108.1
NP_001308037.1
5
867
NM_003615.4
NP_003606.3
5
617
NR_135541.1
5
855
NR_135542.1
5
855
NR_135543.1
5
855
NR_135544.1
5
855
XM_005265598.4
XP_005265655.1
5
1121
XM_005265600.4
XP_005265657.1
5
1122
XM_006713421.2
XP_006713484.1
5
855
XM_011534256.2
XP_011532558.1
5
1128
XM_011534258.1
XP_011532560.1
5
831
XM_011534261.2
XP_011532563.1
5
1132
XM_011534263.2
XP_011532565.1
5
831
XM_011534265.1
XP_011532567.1
5
831
XM_017007526.1
XP_016863015.1
6
752
XM_017007527.1
XP_016863016.1
5
855
GenBank mRNA
AB012130.1
-
2
303
AF047033.1
-
5
617
AF053755.1
-
2
295
AF089726.1
-
5
617
AK309734.1
-
5
855
EU499349.1
-
5
782
EU934246.1
-
5
782
EU934247.1
-
5
794
EU934248.1
-
5
794
EU934249.1
-
5
782
EU934250.1
-
5
794
FJ178574.1
-
5
782
FJ178575.1
-
5
782
FJ178576.1
-
5
782
GU354307.1
-
5
782
GU354308.1
-
5
794
GU354309.1
-
5
794
GU354310.1
-
5
794
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0