Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP2C1 SIRNA Assays ›
Gene Symbol:
ATP2C1
Gene Name:
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases:
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location:
Chr.3: 130850500 - 131016712 on Build GRCh38
UniGene ID:
Hs.584884
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
120526
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP2C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001485.2
NP_001001485.1
13
1497
NM_001001486.1
NP_001001486.1
12
1174
NM_001001487.1
NP_001001487.1
12
1174
NM_001199179.1
NP_001186108.1
13
1403
NM_001199180.1
NP_001186109.1
12
1392
NM_001199181.1
NP_001186110.1
12
1392
NM_001199182.1
NP_001186111.1
11
1275
NM_001199183.1
NP_001186112.1
12
1265
NM_001199184.1
NP_001186113.1
12
1265
NM_001199185.1
NP_001186114.1
12
1316
NM_014382.3
NP_055197.2
12
1316
XM_005247354.2
XP_005247411.1
12
1417
XM_005247355.2
XP_005247412.1
13
1544
XM_005247356.2
XP_005247413.1
13
1297
XM_005247358.2
XP_005247415.1
12
1121
XM_011512686.2
XP_011510988.1
3
226
XM_017006164.1
XP_016861653.1
13
1205
GenBank mRNA
AB037768.1
-
13
1404
AB209265.1
-
1
1164
AF181120.1
-
12
1229
AF181121.1
-
12
1229
AF189723.2
-
12
1148
AF225981.1
-
12
1148
AK074692.1
-
10
855
AK296470.1
-
12
1319
AK299945.1
-
11
1031
AK304175.1
-
12
1121
AK304374.1
-
12
1148
AK314342.1
-
12
1178
AY268374.1
-
12
1123
AY268375.1
-
12
1123
BC028139.1
-
12
1186
5215224
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
cellular calcium ion homeostasis
signal transduction
epidermis development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
actin cytoskeleton reorganization
Golgi calcium ion homeostasis
Golgi calcium ion transport
ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
calcium ion transmembrane transport
signal transducer activity
calcium-transporting ATPase activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-swfkw:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0