Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284338.1 | NP_001271267.1 | 5 | 2168 | ||
NM_001284339.1 | NP_001271268.1 | 5 | 2120 | |||
NM_001284340.1 | NP_001271269.1 | 4 | 2117 | |||
NM_006154.3 | NP_006145.2 | 9 | 802 | |||
NM_198400.3 | NP_940682.2 | 2 | 1952 | |||
NR_104302.1 | 3 | 2055 | ||||
XM_011521624.2 | XP_011519926.1 | 8 | 567 | |||
XM_011521625.2 | XP_011519927.1 | 10 | 740 | |||
XM_011521626.1 | XP_011519928.1 | 8 | 584 | |||
XM_011521627.1 | XP_011519929.1 | 6 | 407 | |||
GenBank mRNA | AK304394.1 | - | 9 | 668 | ||
AK307541.1 | - | 5 | 419 | |||
AL832063.1 | - | 2 | 1953 | |||
AL832359.1 | - | 4 | 640 | |||
AL833293.1 | - | 3 | 1393 | |||
AL833300.1 | - | 5 | 1150 | |||
BC035707.1 | - | 9 | 646 | |||
BC136605.1 | - | 5 | 2145 | 9020595 | ||
BC144285.1 | - | 5 | 2097 | 9052805 | ||
BC152452.1 | - | 9 | 693 | 8862584 | ||
BC152562.1 | - | 9 | 693 | 8860509 | ||
D42055.1 | - | 9 | 693 |