Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000051.3 | NP_000042.3 | 3 | 547 | ||
XM_005271561.4 | XP_005271618.2 | 4 | 608 | |||
XM_005271562.4 | XP_005271619.2 | 3 | 681 | |||
XM_006718843.3 | XP_006718906.1 | 3 | 314 | |||
XM_011542840.2 | XP_011541142.1 | 4 | 402 | |||
XM_011542842.2 | XP_011541144.1 | 3 | 521 | |||
XM_011542843.2 | XP_011541145.1 | 3 | 895 | |||
XM_011542844.2 | XP_011541146.1 | 3 | 519 | |||
XM_017017789.1 | XP_016873278.1 | 4 | 1105 | |||
XM_017017790.1 | XP_016873279.1 | 3 | 502 | |||
XM_017017791.1 | XP_016873280.1 | 3 | 895 | |||
XM_017017792.1 | XP_016873281.1 | 3 | 895 | |||
GenBank mRNA | AB209133.1 | - | 2 | 4685 | ||
BC061584.1 | - | 3 | 350 | |||
U33841.1 | - | 4 | 351 |