Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 42 | 6335 | ||
GenBank mRNA | AB007913.1 | - | 24 | 3296 | ||
AF425231.1 | - | 42 | 6324 | |||
AK126012.1 | - | 21 | 3012 | |||
AK127046.1 | - | 34 | 4942 | |||
AL110281.1 | - | 31 | 4363 | |||
AL117491.1 | - | 32 | 4528 | |||
AL137693.1 | - | 31 | 4341 |