Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 3 | 419 | ||
GenBank mRNA | AF113537.1 | - | 3 | 288 | ||
AF123767.1 | - | 3 | 288 | |||
AF133422.1 | - | 3 | 272 | |||
AK295206.1 | - | 2 | 102 | |||
AK297196.1 | - | 3 | 303 | |||
AK312359.1 | - | 3 | 306 | |||
BC002619.2 | - | 3 | 302 | 3160345 | ||
BC018873.2 | - | 3 | 295 | |||
CR457142.1 | - | 2 | 157 | |||
DC366641.1 | - | 3 | 419 |