Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193329.1 | NP_001180258.1 | 16 | 2619 | ||
NM_032823.5 | NP_116212.3 | 15 | 2405 | |||
XM_005252284.2 | XP_005252341.1 | 14 | 2325 | |||
XM_006717306.1 | XP_006717369.1 | 17 | 2767 | |||
XM_011519119.1 | XP_011517421.1 | 16 | 2622 | |||
XM_011519120.2 | XP_011517422.1 | 15 | 2505 | |||
XM_011519132.1 | XP_011517434.1 | 13 | 1348 | |||
XM_017015229.1 | XP_016870718.1 | 16 | 2665 | |||
XM_017015230.1 | XP_016870719.1 | 16 | 6729 | |||
XM_017015231.1 | XP_016870720.1 | 14 | 6432 | |||
XM_017015233.1 | XP_016870722.1 | 16 | 2652 | |||
XM_017015234.1 | XP_016870723.1 | 15 | 2455 | |||
XM_017015235.1 | XP_016870724.1 | 16 | 2600 | |||
XM_017015236.1 | XP_016870725.1 | 13 | 2158 | |||
XM_017015237.1 | XP_016870726.1 | 13 | 6265 | |||
GenBank mRNA | AF043896.1 | - | 1 | 732 | ||
AF043897.1 | - | 1 | 3465 | |||
BC020194.1 | - | 16 | 1830 | 5020674 | ||
BC029616.1 | - | 15 | 2362 | 3877098 | ||
BX648970.1 | - | 15 | 1705 |