Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145275.1 | NP_001138747.1 | 2 | 746 | ||
NM_003411.3 | NP_003402.2 | 3 | 835 | |||
XM_005262570.2 | XP_005262627.1 | 3 | 918 | |||
XM_011531473.2 | XP_011529775.1 | 2 | 1817 | |||
XM_011531474.2 | XP_011529776.1 | 2 | 1494 | |||
XM_011531475.2 | XP_011529777.1 | 3 | 1908 | |||
XM_017030075.1 | XP_016885564.1 | 3 | 714 | |||
XM_017030076.1 | XP_016885565.1 | 3 | 918 | |||
XM_017030077.1 | XP_016885566.1 | 2 | 1498 | |||
GenBank mRNA | AK310553.1 | - | 3 | 918 | ||
BC114526.1 | - | 2 | 572 | 40054050 | ||
BC114960.1 | - | 3 | 661 | 40054055 | ||
L10393.1 | - | 2 | 547 | |||
M30607.1 | - | 2 | 514 |