Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002011.4 | NP_002002.3 | 9 | 1315 | ||
NM_213647.2 | NP_998812.1 | 9 | 1359 | |||
XM_005265838.3 | XP_005265895.1 | 9 | 1267 | |||
XM_011534464.2 | XP_011532766.1 | 9 | 1429 | |||
XM_011534465.2 | XP_011532767.1 | 9 | 1352 | |||
GenBank mRNA | AB209631.1 | - | 5 | 3436 | ||
AF359246.1 | - | 5 | 636 | |||
AK225280.1 | - | 9 | 1273 | |||
AK225909.1 | - | 9 | 1273 | |||
BC011847.2 | - | 9 | 1345 | 4121396 | ||
JN007471.1 | - | 8 | 1100 | |||
JN007472.1 | - | 8 | 1100 | |||
JN007473.1 | - | 8 | 1100 | |||
JN007474.1 | - | 8 | 1100 | |||
JN007475.1 | - | 8 | 1100 | |||
JN007476.1 | - | 8 | 1100 | |||
JN007477.1 | - | 8 | 1100 | |||
JN007478.1 | - | 8 | 1100 | |||
JN007479.1 | - | 8 | 1100 | |||
JN007480.1 | - | 8 | 1100 | |||
JN007481.1 | - | 8 | 1100 | |||
JN007482.1 | - | 8 | 1100 | |||
L03840.1 | - | 9 | 1270 | |||
X57205.1 | - | 8 | 1155 |