Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 3 | 334 | ||
NM_002823.4 | NP_002814.3 | 3 | 331 | |||
XM_011511518.1 | XP_011509820.1 | 3 | 1064 | |||
XM_011511519.1 | XP_011509821.1 | 3 | 1061 | |||
XM_011511520.1 | XP_011509822.1 | 3 | 963 | |||
XM_011511521.1 | XP_011509823.1 | 3 | 960 | |||
GenBank mRNA | AF348514.1 | - | 3 | 149 | ||
AF452640.1 | - | 3 | 631 | |||
AK223605.1 | - | 3 | 326 | |||
AK314106.1 | - | 3 | 326 | |||
AY169282.1 | - | 3 | 149 | |||
BC003510.1 | - | 3 | 314 | 3610808 | ||
BC022433.1 | - | 3 | 330 | 4277490 | ||
BC034921.1 | - | 3 | 322 | 4335961 | ||
BC051265.1 | - | 3 | 314 | 3832454 | ||
BC066905.1 | - | 3 | 252 | 4830132 | ||
BC070480.1 | - | 3 | 316 | 6205582 | ||
BC071647.1 | - | 3 | 309 | 5112610 | ||
BC071879.1 | - | 3 | 311 | 6425103 | ||
BC098420.1 | - | 3 | 316 | 5017164 | ||
BM839980.1 | - | 3 | 331 | |||
BX647606.1 | - | 1 | 999 | |||
M14483.1 | - | 3 | 281 | |||
M14630.1 | - | 3 | 329 | |||
M14794.1 | - | 3 | 217 | |||
M26708.1 | - | 3 | 304 |