Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBAP2L SIRNA Assays ›
Gene Symbol:
UBAP2L
Gene Name:
ubiquitin associated protein 2 like
Gene Aliases:
NICE-4, NICE4
Chromosome Location:
Chr.1: 154220172 - 154271510 on Build GRCh38
UniGene ID:
Hs.490551
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122722
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBAP2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127320.2
NP_001120792.1
8
774
NM_001287815.1
NP_001274744.1
8
807
NM_001287816.1
NP_001274745.1
8
861
NM_014847.3
NP_055662.3
8
828
XM_005245658.3
XP_005245715.1
8
861
XM_005245667.3
XP_005245724.1
8
861
XM_005245668.3
XP_005245725.1
8
861
XM_005245669.3
XP_005245726.1
8
861
XM_005245670.3
XP_005245727.1
8
861
XM_005245672.3
XP_005245729.1
8
753
XM_005245673.3
XP_005245730.1
8
861
XM_005245674.1
XP_005245731.1
8
753
XM_011510215.2
XP_011508517.1
8
861
XM_011510216.1
XP_011508518.1
8
753
XM_017002972.1
XP_016858461.1
8
886
XM_017002973.1
XP_016858462.1
8
753
XM_017002974.1
XP_016858463.1
8
784
XM_017002975.1
XP_016858464.1
8
861
XM_017002976.1
XP_016858465.1
8
763
XM_017002977.1
XP_016858466.1
8
753
XM_017002978.1
XP_016858467.1
8
784
XM_017002979.1
XP_016858468.1
8
745
XM_017002980.1
XP_016858469.1
8
784
XM_017002981.1
XP_016858470.1
8
745
XM_017002982.1
XP_016858471.1
8
753
XM_017002983.1
XP_016858472.1
8
784
XM_017002984.1
XP_016858473.1
8
784
XM_017002985.1
XP_016858474.1
8
886
XM_017002986.1
XP_016858475.1
8
753
XM_017002987.1
XP_016858476.1
8
784
XM_017002988.1
XP_016858477.1
8
784
XM_017002989.1
XP_016858478.1
8
784
XM_017002990.1
XP_016858479.1
8
745
XM_017002991.1
XP_016858480.1
7
686
GenBank mRNA
AK302662.1
-
8
795
AK302953.1
-
5
522
AK303533.1
-
8
734
BC003170.1
-
7
710
2906083
D63478.1
-
8
767
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
binding of sperm to zona pellucida
hematopoietic stem cell homeostasis
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-lhtlm:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0