Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122822
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
11
1929
NM_001042358.4
NP_001035817.1
10
1787
NM_001042359.3
NP_001035818.1
8
1361
NM_001042360.3
NP_001035819.1
5
1238
NM_001042361.4
NP_001035820.1
8
1236
NM_001042362.3
NP_001035821.1
6
993
NM_001042363.4
NP_001035822.1
10
1772
NM_001042364.4
NP_001035823.1
7
1126
NM_001042365.3
NP_001035824.1
4
785
NM_001320681.1
NP_001307610.1
9
1677
NM_001320682.1
NP_001307611.1
5
883
NM_001320683.1
NP_001307612.1
9
1393
NM_001320684.1
NP_001307613.1
8
1302
NM_001320685.1
NP_001307614.1
10
1849
NM_001320686.1
NP_001307615.1
9
1692
NM_001320688.1
NP_001307617.1
7
1266
NM_001320689.1
NP_001307618.1
6
1034
NM_001320690.1
NP_001307619.1
5
898
NM_001320691.1
NP_001307620.1
8
1601
NM_015605.8
NP_056420.3
9
1378
XM_011539605.2
XP_011537907.1
11
2073
XM_011539606.2
XP_011537908.1
11
3165
XM_011539607.2
XP_011537909.1
11
2064
XM_011539610.2
XP_011537912.1
8
1547
XM_011539611.2
XP_011537913.1
7
1547
XM_017016042.1
XP_016871531.1
11
2010
XM_017016044.1
XP_016871533.1
10
1830
XM_017016045.1
XP_016871534.1
5
3149
XM_017016046.1
XP_016871535.1
10
1692
XM_017016049.1
XP_016871538.1
8
1423
XM_017016050.1
XP_016871539.1
6
1290
XM_017016053.1
XP_016871542.1
8
1266
XM_017016054.1
XP_016871543.1
8
1279
XM_017016055.1
XP_016871544.1
8
1368
XM_017016056.1
XP_016871545.1
7
1274
XM_017016057.1
XP_016871546.1
8
1584
XM_017016058.1
XP_016871547.1
6
1310
XM_017016059.1
XP_016871548.1
7
1268
XM_017016060.1
XP_016871549.1
7
1114
XM_017016061.1
XP_016871550.1
6
1064
XM_017016062.1
XP_016871551.1
6
1184
XM_017016063.1
XP_016871552.1
7
1155
XM_017016064.1
XP_016871553.1
5
907
XM_017016065.1
XP_016871554.1
6
957
GenBank mRNA
AL050040.1
-
9
1635
BX648913.1
-
4
786
CR933668.1
-
5
851
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0