Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145277.1 | NP_001138749.1 | 7 | 1197 | ||
NM_001145278.1 | NP_001138750.1 | 8 | 1195 | |||
NM_018090.4 | NP_060560.1 | 8 | 1273 | |||
XM_011541767.2 | XP_011540069.1 | 8 | 1269 | |||
XM_017001721.1 | XP_016857210.1 | 7 | 1066 | |||
GenBank mRNA | AK021938.1 | - | 4 | 749 | ||
AK023545.1 | - | 7 | 1133 | |||
AK024468.1 | - | 7 | 3825 | |||
AK300764.1 | - | 7 | 1064 | |||
BC017014.1 | - | 8 | 1204 | 3848563 | ||
BC018914.2 | - | 8 | 1207 | 4299890 |