Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173602.2 | NP_775873.2 | 22, 23 | 2813 | ||
XM_005269044.1 | XP_005269101.1 | 22, 23 | 2816 | |||
XM_006719520.3 | XP_006719583.1 | 23, 24 | 2789 | |||
XM_006719521.3 | XP_006719584.1 | 22, 23 | 2626 | |||
XM_011538594.2 | XP_011536896.1 | 23, 24 | 2842 | |||
XM_011538595.2 | XP_011536897.1 | 21, 22 | 2674 | |||
XM_011538596.2 | XP_011536898.1 | 22, 23 | 2629 | |||
XM_017019699.1 | XP_016875188.1 | 23, 24 | 2824 | |||
XM_017019700.1 | XP_016875189.1 | 22, 23 | 2798 | |||
XM_017019701.1 | XP_016875190.1 | 22, 23 | 2795 | |||
XM_017019702.1 | XP_016875191.1 | 20, 21 | 2645 | |||
XM_017019703.1 | XP_016875192.1 | 21, 22 | 2592 | |||
XM_017019704.1 | XP_016875193.1 | 21, 22 | 2600 | |||
XM_017019705.1 | XP_016875194.1 | 21, 22 | 2582 | |||
GenBank mRNA | AB040896.2 | - | 13, 14 | 1427 | ||
AK097369.1 | - | 4, 5 | 369 | |||
AK126758.1 | - | 11, 12 | 1184 | |||
BC007671.2 | - | 6, 7 | 737 | |||
BC030156.2 | - | 6, 7 | 737 |